Dimethyl glyoxime, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime
M-Clarity™ quality level = MQ100
03858-100G
03858-500G
SIAL03858-100GEA
101
GBP
SIAL03858-100G
SIAL03858-500G
Dimethyl glyoxime, Sigma-Aldrich®
Dimethyl glyoxime
Formula:
CH₃C(=NOH)C(=NOH)CH₃ Boiling Pt: 53 °C (8 mmHg) Melting Pt: 240…241 °C Density: 1.37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002117 CAS Number: 95-45-4 EINECS: 202-420-1 Merck Index: 13,03272 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...