Urethane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Carbamic acid ethyl ester, Ethyl carbamate
94300-1KG
94300-250G
94300-50G
SIAL94300-1KGEA
385
GBP
SIAL94300-1KG
SIAL94300-250G
SIAL94300-50G
Urethane, Sigma-Aldrich®
Urethane
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 182 °C (1013 hPa) Melting Pt: 48…50 °C Density: 1.11 g/cm³ (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00007966 CAS Number: 51-79-6 EINECS: 200-123-1 UN: 2757 ADR: 6.1,II Merck Index: 12,10013 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...