Ethyl palmitate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Hexadecanoic acid ethyl ester
, Ethyl hexadecanoate
, Hexadecylic acid ethyl ester
, Palmitic acid ethyl ester
M-Clarity™ quality level = MQ100
SIALW245100-1KG-KEA
290
GBP
SIALW245100-1KG-K
SIALW245100-250G-K
SIALW245100-5KG-K
SIALW245100-10KG-K
Ethyl palmitate, Sigma-Aldrich®
Ethyl palmitate
Formula:
C₁₈H₃₆O₂ MW: 284.48 g/mol Boiling Pt: 342 °C (1013 hPa) Melting Pt: 24 °C Density: 0.86 g/cm³ (20 °C) |
MDL Number:
MFCD00008996 CAS Number: 628-97-7 EINECS: 211-064-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...