2-Methylbutyl acetate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Acetic acid 2-Methylbutyl ester
M-Clarity™ quality level = MQ300
W364428-100G-K
W364428-1KG-K
W364428-4KG-K
SIALW364428-100G-KEA
86
GBP
SIALW364428-100G-K
SIALW364428-1KG-K
SIALW364428-4KG-K
2-Methylbutyl acetate, Sigma-Aldrich®
2-Methylbutyl acetate
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 140…142 °C (1013 hPa) Density: 0.876 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00040494 CAS Number: 624-41-9 EINECS: 210-843-8 UN: 1104 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...