Ethyl decanoate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester
M-Clarity™ quality level = MQ300
W243213-100G-K
W243213-1KG-K
W243213-4KG-K
SIALW243213-100G-KEA
76.5
GBP
SIALW243213-100G-K
SIALW243213-1KG-K
SIALW243213-4KG-K
Ethyl decanoate, Sigma-Aldrich®
Ethyl decanoate
Formula:
C₁₂H₂₄O₂ MW: 200.32 g/mol Boiling Pt: 245 Melting Pt: –20 °C Density: 0.862 g/cm³ (20 °C) |
MDL Number:
MFCD00009581 CAS Number: 110-38-3 EINECS: 203-761-9 Merck Index: 13,03811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...