Propyl butyrate
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ300
W293415-100G-K
W293415-1KG-K
W293415-25G-K
W293415-4KG-K
SIALW293415-100G-KEA
128
GBP
SIALW293415-100G-K
SIALW293415-1KG-K
SIALW293415-25G-K
SIALW293415-4KG-K
Propyl butyrate
Propyl butyrate
Formula:
C₇H₁₄O₂ |
MDL Number:
MFCD00009396 CAS Number: 105-66-8 EINECS: 203-320-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...