Trilaurin (glycerol trilaurate), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Glyceryl tridodecanoate

M-Clarity™ quality level = MQ200

T4891-5G T4891-100MG
SIALT4891-5GEA 183 GBP
SIALT4891-5G SIALT4891-100MG
Trilaurin (glycerol trilaurate), Sigma-Aldrich®
Trilaurin (Glycerol trilaurate)
Formula: C₃₉H₇₄O₆
Boiling Pt: 643 °C (1013 hPa)
Melting Pt: 49 °C
Density: 0.9713 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00026559
CAS Number: 538-24-9
EINECS: 208-687-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR