Trichloroacetyl chloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
151599-100G
151599-25G
151599-500G
SIAL151599-100GEA
134
GBP
SIAL151599-100G
SIAL151599-25G
SIAL151599-500G
Trichloroacetyl chloride, Sigma-Aldrich®
Trichloroacetyl chloride
Formula:
C₂Cl₄O MW: 181.83 g/mol Boiling Pt: 118 °C (1013 hPa) Melting Pt: –57 °C Density: 1.62 g/cm³ (20 °C) |
MDL Number:
MFCD00000792 CAS Number: 76-02-8 EINECS: 200-926-7 UN: 2442 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...