(S)-(-)-1-Phenylethyl isocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-(-)-α-Phenethyl isocyanate, (S)-(-)-α-Methylbenzyl isocyanate, (S)-1-phenylethylisocyanate, (S)-(-)-1-Phenethyl isocyanate

013634-1G 013634-5G
FLUO013634-1GEA 0 GBP
FLUO013634-1G FLUO013634-5G
(S)-(-)-1-Phenylethyl isocyanate
(S)-(-)-1-Phenylethyl isocyanate
Formula: C₉H₉NO
Boiling Pt: 55 °C (1013 hPa)
Density: 1,05 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002036
CAS Number: 14649-03-7
EINECS: 238-698-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR