(R)-(+)-1-Phenylethyl isocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013633-1G
013633-5G
FLUO013633-1GEA
0
GBP
FLUO013633-1G
FLUO013633-5G
(R)-(+)-1-Phenylethyl isocyanate
(R)-(+)-1-Phenylethyl isocyanate
Formula:
C₉H₉NO Boiling Pt: 74…76 °C (7 hPa) Density: 1,05 g/cm³ (20 °C) Flash Pt: 65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06739387 CAS Number: 33375-06-3 EINECS: 251-485-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...