Iron(III) fluoride trihydrate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006157-100G 006157-500G 006157-1KG
FLUO006157-100GEA 0 GBP
FLUO006157-100G FLUO006157-500G FLUO006157-1KG
Iron(III) fluoride trihydrate
Iron (III) fluoride trihydrate
Formula: F₃Fe·3H₂O
Density: 2.3 g/cm³ (20 °C)
MDL Number: MFCD00149718
CAS Number: 15469-38-2
EINECS: 232-002-4
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR