(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
M-Clarity™ quality level = MQ100
295817-1G
295817-250MG
295817-5G
SIAL295817-1GEA
115
GBP
SIAL295817-1G
SIAL295817-250MG
SIAL295817-5G
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula:
C₄₄H₃₂P₂ MW: 622.69 g/mol Melting Pt: 239…242 °C |
MDL Number:
MFCD00010805 CAS Number: 76189-55-4 Merck Index: 13,01223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...