Europium trinitrate pentahydrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Europium (III) nitrate pentahydrate
M-Clarity™ quality level = MQ100
207918-10G
207918-1G
207918-50G
SIAL207918-10GEA
613
GBP
SIAL207918-10G
SIAL207918-1G
SIAL207918-50G
Europium trinitrate pentahydrate, Sigma-Aldrich®
Europium trinitrate pentahydrate
Formula:
EuN₃O₉·5H₂O MW: 428.05 |
CAS Number:
63026-01-7 EINECS: 233-380-3 UN: 1477 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...