Hydrochloric acid 36,5%, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ200

H1758-100ML H1758-500ML
SIALH1758-100MLEA 119 GBP
SIALH1758-100ML SIALH1758-500ML
Hydrochloric acid 36,5%, Sigma-Aldrich®
Hydrochloric acid
Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR