Yeast viability kit for staining of live/dead yeast cells with AO/PI, ViaStain®
Supplier: Nexcelom Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NEXCCSK-0102-10MLEA 884 GBP
NEXCCSK-0102-10ML NEXCCS1-0109-5ML NEXCCSK-0102-2ML
Yeast viability kit for staining of live/dead yeast cells with AO/PI, ViaStain®
Assays Cellular Assays
Determining yeast viability and concentration during biofuel and brewing production is essential for process optimisation and monitoring of alcohol production.
Many of these complex yeast samples increase the difficulty for automated counting using bright field method as well as manual counting due to the debris particles in the sample. By using a combination of fluorescent dyes acridine orange (AO) and propidium iodide (PI) Cellometer instruments can accurately determine the concentration and viability of the complex samples.
Acridine orange (AO) and propidium iodide (PI) are nuclear staining (nucleic acid binding) dyes. AO is permeable to both live and dead cells and stains all nucleated cells to generate green fluorescence. PI enters dead cells with compromised membranes and stains all dead nucleated cells to generate red fluorescence. Cells stained with both AO and PI fluoresce red due to quenching, so all live nucleated cells fluoresce green and all dead nucleated cells fluoresce red.