ESD safety shoes, lace-up, uvex 2 trend, 6907
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6907.7/38 6907.7/39 6907.7/40 6907.7/41 6907.7/42 6907.7/43 6907.7/44 6907.7/45 6907.7/46 6907.7/47 6907.7/48 6907.7/49 6907.7/50 6907.7/51 6907.7/52 6907.8/38 6907.8/39 6907.8/40 6907.8/41 6907.8/42 6907.8/43 6907.8/44 6907.8/45 6907.8/46 6907.8/47 6907.8/48 6907.8/49 6907.8/50 6907.8/51 6907.8/52 6907.9/38 6907.9/39 6907.9/40 6907.9/41 6907.9/42 6907.9/43 6907.9/44 6907.9/45 6907.9/46 6907.9/47 6907.9/48 6907.9/49 6907.9/50 6907.9/51 6907.9/52
UVEX6907.7/38EA 0 GBP
UVEX6907.7/38 UVEX6907.7/39 UVEX6907.7/40 UVEX6907.7/41 UVEX6907.7/42 UVEX6907.7/43 UVEX6907.7/44 UVEX6907.7/45 UVEX6907.7/46 UVEX6907.7/47 UVEX6907.7/48 UVEX6907.7/49 UVEX6907.7/50 UVEX6907.7/51 UVEX6907.7/52 UVEX6907.8/38 UVEX6907.8/39 UVEX6907.8/40 UVEX6907.8/41 UVEX6907.8/42 UVEX6907.8/43 UVEX6907.8/44 UVEX6907.8/45 UVEX6907.8/46 UVEX6907.8/47 UVEX6907.8/48 UVEX6907.8/49 UVEX6907.8/50 UVEX6907.8/51 UVEX6907.8/52 UVEX6907.9/38 UVEX6907.9/39 UVEX6907.9/40 UVEX6907.9/41 UVEX6907.9/42 UVEX6907.9/43 UVEX6907.9/44 UVEX6907.9/45 UVEX6907.9/46 UVEX6907.9/47 UVEX6907.9/48 UVEX6907.9/49 UVEX6907.9/50 UVEX6907.9/51 UVEX6907.9/52
ESD safety shoes, lace-up, uvex 2 trend, 6907
Shoes and Boots
ESD sporty S1 safety shoe for men with wide fit and slip resistance PU sole.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable, perforated microvelours
  • Steel toe cap
  • Reflective details for enhanced visibility
  • Excellent shock absorption
  • Uvex climazone foot climate management, uvex 1 climate comfort insole, uvex multi-width system helps maintain foot temperature for optimum wearer comfort
Safety Shoes feature an ergonomically designed outsole with excellent slip resistance, as well as a stabilising uvex anti-twist heelMore Product Information