Aristolochic acid ≥98%, crystalline solid

Supplier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

21520-100 21520-25 21520-10 21520-50
CAYM21520-100EA 497 GBP
CAYM21520-100 CAYM21520-25 CAYM21520-10 CAYM21520-50
Aristolochic acid ≥98%, crystalline solid
Aristolochic acid

Aristolochic acid is an alkaloid originally isolated from A. radix that inhibits inflammatory phospholipase A₂ (PLA₂) activity (IC₅₀=40 µM in human neutrophils).


It inhibits human synovial fluid PLA₂-induced edema in a dose-dependent manner in vivo which positively correlates with in vitro inhibition of PLA₂. Aristolochic acid inhibits hemolytic activity of PLA₂s TFV PL-X, TFV PL-Ia, and TFV PL-Ib from T. flavoviridis venom (IC₅₀s=4.6, 4.0, and 3.9 µM, respectively) but enhances the edema-inducing activity of TFV PL-Ia and TFV PL-Ib. Aristolochic acid exposure is also associated with progressive interstitial fibrosing nephropathy and upper urinary tract urothelial carcinomas.

Formula: C₁₇H₁₁NO₇
MW: 341.275 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238539
CAS Number: 313-67-7
UN: 1544
ADR: 6.1 III

Order Now

Specification Test Results

Purity ≥98%
Formulation A Crystalline Solid
λmax 224, 320, 388 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR