ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1 Trainer, 9559
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9559.7/38 9559.7/39 9559.7/40 9559.7/41 9559.7/42 9559.7/43 9559.7/44 9559.7/45 9559.7/46 9559.7/47 9559.7/48 9559.7/49 9559.7/50 9559.7/51 9559.7/52 9559.8/38 9559.8/39 9559.8/40 9559.8/41 9559.8/42 9559.8/43 9559.8/44 9559.8/45 9559.8/46 9559.8/47 9559.8/48 9559.8/49 9559.8/50 9559.8/51 9559.8/52 9559.9/38 9559.9/39 9559.9/40 9559.9/41 9559.9/42 9559.9/43 9559.9/44 9559.9/45 9559.9/46 9559.9/47 9559.9/48 9559.9/49 9559.9/50 9559.9/51 9559.9/52
UVEX9559.7/38EA 0 GBP
UVEX9559.7/38 UVEX9559.7/39 UVEX9559.7/40 UVEX9559.7/41 UVEX9559.7/42 UVEX9559.7/43 UVEX9559.7/44 UVEX9559.7/45 UVEX9559.7/46 UVEX9559.7/47 UVEX9559.7/48 UVEX9559.7/49 UVEX9559.7/50 UVEX9559.7/51 UVEX9559.7/52 UVEX9559.8/38 UVEX9559.8/39 UVEX9559.8/40 UVEX9559.8/41 UVEX9559.8/42 UVEX9559.8/43 UVEX9559.8/44 UVEX9559.8/45 UVEX9559.8/46 UVEX9559.8/47 UVEX9559.8/48 UVEX9559.8/49 UVEX9559.8/50 UVEX9559.8/51 UVEX9559.8/52 UVEX9559.9/38 UVEX9559.9/39 UVEX9559.9/40 UVEX9559.9/41 UVEX9559.9/42 UVEX9559.9/43 UVEX9559.9/44 UVEX9559.9/45 UVEX9559.9/46 UVEX9559.9/47 UVEX9559.9/48 UVEX9559.9/49 UVEX9559.9/50 UVEX9559.9/51 UVEX9559.9/52
ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1 Trainer, 9559
Shoes and Boots
Metal-free, lightweight, improved fit and optimum cushioning, padding significantly increase comfort, made of micro velour upper, soft padded collar and outsole made from dual-density PUR sole with excellent slip resistance. Sole materials are free from silicones, plasticisers and other paint-wetting impairment substances.
  • Climazone technology for improved comfort
  • Double individually adjustable velcro fastening
  • Breathable textile lining
  • Removable anti-static comfort insole with moisture control system; with a moisture-wicking system
  • Anti-slip sole with shock absorption at heel and front of the foot
  • 100% metal-free xenova® toecap, compact design, anatomical shape, good lateral stability, no thermal conductivity
  • Fulfils ESD requirements: resistance to ground inferior to 35 megaohms
  • Suitable for indoor and outdoor works
Contains uvex xenova® plastic cap which is the safety feature of this shoes. Suitable for people allergic to chrome, made from synthetic materials.
Colour: Black/green
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC