Welding spectacles, uvex futura
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9180.143 9180.145
UVEX9180.143EA 0 GBP
UVEX9180.143 UVEX9180.145
Welding spectacles, uvex futura
Safety Glasses
These classic welding protection feature new filter technology for UV and IR radiation protection.
  • Perfect colour recognition thanks to uvex sunglare filters
  • Minimises damage from welding sparks
  • Multi-level side arm inclination and four different side arm lengths adjust to suit any facial shape
  • Soft duo-flex ear pieces for a pressure free fit
Lens coating of uvex infradur with grey welding protection tinting.
Certifications: EN 166, EN 169