Solvent extraction system, MonoTrap®
Supplier: GL Sciences B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1050-71101 1050-71201 1050-72101 1050-72201
GLSC1050-71101EA 426 GBP
GLSC1050-71101 GLSC1050-71201 GLSC1050-72101 GLSC1050-72201
Solvent extraction system, MonoTrap®
Protein Purification Protein Purification Resins
This extraction media is designed for simple and rapid enrichment of flavors, aromas, and fragrances, and can be easily used for the analysis of volatile and semi-volatile compounds for quality control, environmental and forensic applications.
  • Easy to use
  • High efficiency adsorption
  • Complete desorption with low solvent volume
  • Hydrophobic surface