3-(Piperidin-2-yl)pyridine ≥97% (by titrimetric analysis), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (±) Anabasine, 2-Pyridin-3-ylpiperidine

M-Clarity™ quality level = MQ100

284599-100MG 284599-1G
SIAL284599-100MGEA 182 GBP
SIAL284599-100MG SIAL284599-1G
3-(Piperidin-2-yl)pyridine ≥97% (by titrimetric analysis), Sigma-Aldrich®
3-(Piperidin-2-yl)pyridine

Anabasine, a tobacco alkaloid, inhibited androstenedione conversion to estrogen in a dose-dependent manner.


Used as a biomarker of active tobacco use.

Formula: C₁₀H₁₄N₂
MW: 162.236 g/mol
Boiling Pt: 96 °C/12 mmHg (lit.)
Density: 1.046 g/ml (25 °C(lit.)) (lit.)
Flash Pt: 93 °C
MDL Number: MFCD00006370
CAS Number: 13078-04-1
UN: 3140
ADR: 6.1,III

Order Now

Specification Test Results

Appearance (Colour) Colourless to Yellow
Appearance (Form) Liquid
Infrared Spectrum Conforms to Structure
Purity (Titration by HCl04) ≥97.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR