Kromasil® Classic HPLC Columns, 60 Å
Supplier: HICHROM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S05CNA15 S05CNA25 S05DIA25 S05DIC15 S05DINGC S05HDA15 S05HDC05 S05HDNGC S05SIC15 S05SIF10 S05SINGC S05CNC05 S05DIC05 S10CNR25 S10CN025
HISMS05CNA15EA 433 GBP
HISMS05CNA15 HISMS05CNA25 HISMS05DIA25 HISMS05DIC15 HISMS05DINGC HISMS05HDA15 HISMS05HDC05 HISMS05HDNGC HISMS05SIC15 HISMS05SIF10 HISMS05SINGC HISMS05CNC05 HISMS05DIC05 HISMS10CNR25 HISMS10CN025
Kromasil® Classic HPLC Columns, 60 Å
Chromatography Columns
The Kromasil® classic 60 Å family of products is the choice for small, organic molecules when a large, accessible surface area is key for separating peaks in analysis. Derivatised stationary phase materials based on Kromasil® 60 Å silica are developed and manufactured to give high reproducibility and chemical stability. Scientists can benefit from this range of products for applications within normal phase, reversed phase, HILIC and SFC.
  • Spherical porous high purity silica
  • Small pore size and high surface area silica for high loadability
  • Analytical to
More Product Information