Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm

178810-10L 178810-1L 178810-2.5L 178810-20L
178810-10LEA 416 GBP
178810-10L 178810-1L 178810-2.5L 178810-20L
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Tetrahydrofuran

  • ≥95% for general laboratory use.

Formula: C₄H₈O
Boiling Pt: 65 °C (1.013 hPa)
Melting Pt: –108 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Assay BHT (GC) 200 - 300 ppm
APHA Max. 20
Refractive Index n20/D 1.407 (lit.)
Explosion Limit 11.8 %
Vapor Pressure 114 mmHg ( 15 °C)
Vapor Density 2.5 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR