Human N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S7388-100MG S7388-250MG S7388-25MG S7388-500MG
SIALS7388-100MGEA 476 GBP
SIALS7388-100MG SIALS7388-250MG SIALS7388-25MG SIALS7388-500MG
Human N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide
Proteins and Peptides
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide is a colorimetric substrate for chymotrypsin, human leukocyte cathepsin G and peptidyl prolyl isomerase.