Cable ties, non-releasable, metal content
Supplier: Azpack
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
129-1128 129-1129 129-1130
129-1128EA 108 GBP
129-1128 129-1129 129-1130
Cable ties, non-releasable, metal content
Ties
These polyamide cable ties are designed for the food and beverage processing industries to meet the safety standards set by Hazard Analysis and Critical Control Points (HACCP).
  • Metal content in head and strap for identification by metal detectors and X-ray machines
  • Blue color for visual detection reduces the risk of contamination
  • Operating temperature from −40 to +85 °C
They are ideally suited for the management of cables and tubing in processing and packaging areas.
Certifications: Self extinguishing class UL94 HB.
Order Now
SPECIFICATIONS
Environmentally Preferable