N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥95.0% (by HPLC, sum of enantiomers), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Fmoc-Cys(Trt)-OH, N-Fmoc-S-trityl-L-cysteine

47695-5G 47695-100G 47695-1KG
SIAL47695-5GEA 83.5 GBP
SIAL47695-5G SIAL47695-100G SIAL47695-1KG
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥95.0% (by HPLC, sum of enantiomers), Sigma-Aldrich®
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine
Formula: C₃₇H₃₁NO₄S
MW: 585.72 g/mol
Melting Pt: 170…173 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00038538
CAS Number: 103213-32-7

Order Now

Specification Test Results

Assay (HPLC area %) ≥95.0% rel
Specific rotation (20/D) 16 ±2°
Concentration C=1 in THF
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR