Sulphuric acid ≥98%

Supplier: DISTRIBUTOR
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

SU6071L SU6072
DIUKSU6071LEA 32.3 GBP
DIUKSU6071L DIUKSU6072
Sulphuric acid ≥98%
Sulphuric acid
Formula: H₂SO₄
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Purity 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR