2,5-Dimethylthiophene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D188603-100G
D188603-25G
SIALD188603-100GEA
249
GBP
SIALD188603-100G
SIALD188603-25G
2,5-Dimethylthiophene, Sigma-Aldrich®
2,5-Dimethylthiophene
Formula:
C₆H₈S MW: 112.2 g/mol Boiling Pt: 134 °C (1013 hPa) Melting Pt: –63 °C Density: 0.984 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005452 CAS Number: 638-02-8 EINECS: 211-313-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...