2-Acetylthiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

005042-500G 005042-100G
FLUO005042-500GEA 0 GBP
FLUO005042-500G FLUO005042-100G
2-Acetylthiophene
2-Acetylthiophene
Formula: C₆H₆OS
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.1704 g/cm³ (20 °C)
MDL Number: MFCD00005442
CAS Number: 88-15-3
EINECS: 201-804-6
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR