2-Bromo-3-methylthiophene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

337021-1G 337021-5G
SIAL337021-1GEA 52 GBP
SIAL337021-1G SIAL337021-5G
2-Bromo-3-methylthiophene, Sigma-Aldrich®
2-Bromo-3-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 14…16 °C (0.3 mmHg)
Density: 1.5844 g/cm³ (17.5 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00059741
CAS Number: 14282-76-9
EINECS: 238-175-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR