Hexamethyldisiloxane, 0.65 cSt (25°C), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
52630-100ML
52630-500ML
SIAL52630-100MLEA
58.2
GBP
SIAL52630-100ML
SIAL52630-500ML
Hexamethyldisiloxane, 0.65 cSt (25°C), Sigma-Aldrich®
Hexamethyldisiloxane
Formula:
C₆H₁₈OSi₂ Boiling Pt: 101 °C (1013 hPa) Melting Pt: –59 °C Density: 0.764 g/cm³ (20 °C) Flash Pt: 0.6 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008265 CAS Number: 107-46-0 EINECS: 203-492-7 UN: 1993 ADR: 3,II REACH: 01-2119496108-31 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...