2-Acetyl-1-methyl-1H-pyrrole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole
002488-5G
002488-25G
002488-25ML
002488-5ML
FLUO002488-5GEA
0
GBP
FLUO002488-5G
FLUO002488-25G
FLUO002488-25ML
FLUO002488-5ML
2-Acetyl-1-methyl-1H-pyrrole
2-Acetyl-1-methyl-1H-pyrrole
Formula:
C₇H₉NO Boiling Pt: 200 °C (1013 hPa) Density: 1,0445 g/cm³ (15 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003089 CAS Number: 932-16-1 EINECS: 213-247-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...