2-Acetyl-1H-pyrrole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002744-5G 002744-25G
FLUO002744-5GEA 0 GBP
FLUO002744-5G FLUO002744-25G
2-Acetyl-1H-pyrrole
2-Acetyl-1H-pyrrole
Formula: C₆H₇NO
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 63…65 °C
MDL Number: MFCD00005220
CAS Number: 1072-83-9
EINECS: 214-016-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR