4-tert-Butylpyridine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-(tert-Butyl)pyridine
M-Clarity™ quality level = MQ100
142379-100G
142379-25G
SIAL142379-100GEA
579
GBP
SIAL142379-100G
SIAL142379-25G
4-tert-Butylpyridine, Sigma-Aldrich®
4-tert-Butylpyridine
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: –0.9 °C Density: 0.9219 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006435 CAS Number: 3978-81-2 EINECS: 223-614-2 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...