2-Picoline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Methylpyridine, α-Picoline

M-Clarity™ quality level = MQ200

109835-1L 109835-25ML 109835-500ML
SIAL109835-1LEA 120 GBP
SIAL109835-1L SIAL109835-25ML SIAL109835-500ML
2-Picoline, Sigma-Aldrich®
2-Picoline
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 129.5 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.944 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00006332
CAS Number: 109-06-8
EINECS: 203-643-7
UN: 2313
ADR: 3,III
Merck Index: 12,07554

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR