1-Methyl-2-pyridone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Methylpyridin-2-ol, N-Methylpyridin-2-ol, 1-Methylpyridin-2(1H)-one, N-Methyl-2-pyridone
M-Clarity™ quality level = MQ100
M78259-100G
M78259-25G
SIALM78259-100GEA
533
GBP
SIALM78259-100G
SIALM78259-25G
1-Methyl-2-pyridone, Sigma-Aldrich®
1-Methyl-2-pyridone
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 250 Melting Pt: 33 Density: 1.1321 g/cm³ (18 °C) |
MDL Number:
MFCD00006520 CAS Number: 694-85-9 EINECS: 211-777-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...