2-(2-Aminoethyl)pyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine
013702-5G
013702-1G
FLUO013702-5GEA
0
GBP
FLUO013702-5G
FLUO013702-1G
2-(2-Aminoethyl)pyridine
2-(2-Aminoethyl)pyridine
Formula:
C₇H₁₀N₂ Boiling Pt: 49…50 °C (0.03 mmHg) Density: 1,02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006367 CAS Number: 2706-56-1 EINECS: 220-295-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...