2-Fluoro-5-methylpyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Fluoro-5-picoline

008920-5G 008920-25G
FLUO008920-5GEA 0 GBP
FLUO008920-5G FLUO008920-25G
2-Fluoro-5-methylpyridine
2-Fluoro-5-methylpyridine
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 152…155 °C (1013 hPa)
Density: 1.077 g/cm³ (20 °C)
MDL Number: MFCD00041225
CAS Number: 2369-19-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR