2-Fluoro-4-methylpyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Fluoro-4-picoline

006287-5G 006287-25G
FLUO006287-5GEA 0 GBP
FLUO006287-5G FLUO006287-25G
2-Fluoro-4-methylpyridine
2-Fluoro-4-methylpyridine
Formula: C₆H₆FN
Boiling Pt: 157 °C (1013 hPa)
Density: 1.0805 g/cm³ (25 °C)
MDL Number: MFCD00041224
CAS Number: 461-87-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR