2-Chloro-4-methylpyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

010404-5G 010404-25G 010404-100G
FLUO010404-5GEA 0 GBP
FLUO010404-5G FLUO010404-25G FLUO010404-100G
2-Chloro-4-methylpyridine
2-Chloro-4-methylpyridine
Formula: C₆H₆ClN
Boiling Pt: 194…195 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1,456 g/cm³ (25 °C)
MDL Number: MFCD00023418
CAS Number: 3678-62-4
EINECS: 222-951-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR