2-Amino-5-fluoropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005231-1G 005231-5G 005231-25G
FLUO005231-1GEA 0 GBP
FLUO005231-1G FLUO005231-5G FLUO005231-25G
2-Amino-5-fluoropyridine
2-Amino-5-fluoropyridine
Formula: C₅H₅FN₂
MDL Number: MFCD01861120
CAS Number: 21717-96-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR