2-Hydroxynicotinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Hydroxypyridine-3-carboxylic acid, 1,2-Dihydro-2-oxonicotinic acid, 2-Hydroxy-3-pyridinecarboxylic acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic acid

009264-100G 009264-500G
FLUO009264-100GEA 0 GBP
FLUO009264-100G FLUO009264-500G
2-Hydroxynicotinic acid
2-Hydroxynicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00010100
CAS Number: 609-71-2
EINECS: 210-198-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR