Nicotinic acid (Niacin)

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3

3815.1 3815.2
ROTH3815.1EA 0 GBP
ROTH3815.1 ROTH3815.2
Nicotinic acid (Niacin)
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR