4-Chloro-2-hydroxypyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

011626-500MG 011626-5G
FLUO011626-500MGEA 0 GBP
FLUO011626-500MG FLUO011626-5G
4-Chloro-2-hydroxypyridine
4-Chloro-2-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.55 g/mol
MDL Number: MFCD03093022
CAS Number: 40673-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR