2-Chloro-5-nitropyridine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
C61801-25G
C61801-5G
SIALC61801-25GEA
119
GBP
SIALC61801-25G
SIALC61801-5G
2-Chloro-5-nitropyridine, Sigma-Aldrich®
2-Chloro-5-nitropyridine
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Melting Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006240 CAS Number: 4548-45-2 EINECS: 224-908-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...