5-Amino-2-chloropyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Chloro-5-pyridylamine, 6-Chloropyridin-3-amine, 2-Chloropyridin-5-amine
010694-25G
010694-100G
FLUO010694-25GEA
0
GBP
FLUO010694-25G
FLUO010694-100G
5-Amino-2-chloropyridine
5-Amino-2-chloropyridine
Formula:
C₅H₅ClN₂ Boiling Pt: 225 °C (1013 hPa) Melting Pt: 83 °C |
MDL Number:
MFCD00006243 CAS Number: 5350-93-6 EINECS: 226-322-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...