3-Amino-2,5-dichloropyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
017241-2.5G
017241-1G
FLUO017241-2.5GEA
0
GBP
FLUO017241-2.5G
FLUO017241-1G
3-Amino-2,5-dichloropyridine
3-Amino-2,5-dichloropyridine
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00204157 CAS Number: 78607-32-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...