2-Chloro-4-iodopyridine ≥99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

L19623.06 L19623.03
L19623.06EA 131 GBP
L19623.06 L19623.03
2-Chloro-4-iodopyridine ≥99%
2-Chloro-4-iodopyridine

Purity: 99% CAS No.: 153034-86-7 Molecular Formula: C5H3ClIN Formula Weight: 239.44 Melting Point: 42-44° Beilstein Reference: 108668

Formula: C₅H₃ClIN
MW: 239.44 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD01861983
CAS Number: 153034-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR