2-Chloronicotinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

010748-250G 010748-50G
FLUO010748-250GEA 0 GBP
FLUO010748-250G FLUO010748-50G
2-Chloronicotinic acid
2-Chloronicotinic acid
Formula: C₆H₄ClNO₂
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR