6-Chloronicotinamide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Chloropyridine-3-carboxamide
M-Clarity™ quality level = MQ100
C58002-10G
C58002-1G
SIALC58002-10GEA
137
GBP
SIALC58002-10G
SIALC58002-1G
6-Chloronicotinamide, Sigma-Aldrich®
6-Chloronicotinamide
Formula:
C₆H₅ClN₂O MW: 156.57 g/mol |
MDL Number:
MFCD00006242 CAS Number: 6271-78-9 EINECS: 228-456-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...